missing translation for 'onlineSavingsMsg'
Learn More

Phloretin, MedChemExpress

Product Code. 16479895
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479895 250 mg 250mg
16412356 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479895 Supplier Medchem Express Supplier No. HYN0142
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phloretin (NSC 407292; RJC 02792) is a flavonoid extracted from Prunus mandshurica, has anti-inflammatory activities.

Phloridzin is a specific, competitive and orally active inhibitor of sodium/glucose cotransporters in the intestine (SGLT1) and kidney (SGLT2). Phloretin inhibits Yeast-made GLUT1 as well as Human erythrocyte GLUT1 with IC50values of 49 μM and 61 μM, respectively.Phloretin has the potential for the treatment of rheumatoid arthritis (RA) and allergic airway inflammation.

Specifications

Chemical Name or Material Phloretin
CAS 60-82-2
Color Pink
Purity Grade Notes Research
Molecular Formula C15H14O5
Quantity 250 mg
Synonym NSC 407292 RJC 02792
Solubility Information DMSO : 50 mg/mL (182.30 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(O)C=C(O)C=C1O)CCC2=CC=C(O)C=C2
Molecular Weight (g/mol) 274.27
Formula Weight 274.27
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.