missing translation for 'onlineSavingsMsg'
Learn More

Phloracetophenone, MedChemExpress

Product Code. 16489326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489326 Supplier Medchem Express Supplier No. HYW008226
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phloracetophenone (2,4,6-trihydroxyacetophenone) is the aglycone part of acetophenone glycoside obtained from Curcuma comosa Roxb, with cholesterol-lowering activity. Phloracetophenone enhances cholesterol 7α-hydroxylase (CYP7A1) activity. Phloracetophenone stimulats bile secretion mediated through Mrp2.

Phloracetophenone (2,4,6-trihydroxyacetophenone) is the aglycone part of acetophenone glycoside obtained from Curcuma comosa Roxb, with cholesterol-lowering activity. Phloracetophenone enhances cholesterol 7α-hydroxylase (CYP7A1) activity. Phloracetophenone stimulats bile secretion mediated through Mrp2.

Specifications

Chemical Name or Material Phloracetophenone
CAS 480-66-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C8H8O4
Quantity 100 mg
Synonym 2,4,6-trihydroxyacetophenone 1-(2,4,6-Trihydroxyphenyl)ethanone
Solubility Information DMSO : ≥ 100 mg/mL (594.71 mM) ∣H2O : 2 mg/mL (11.89 mM; Need ultrasonic)
SMILES CC(=O)C1=C(O)C=C(O)C=C1O
Molecular Weight (g/mol) 168.15
Formula Weight 168.15
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.