missing translation for 'onlineSavingsMsg'
Learn More

Phleomycin, Fisher BioReagents

Product Code. 10319787
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10319787 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10319787 Supplier Fisher BioReagents Supplier No. BP2962100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Disrupts the integrity of DNA, and blocks S-phase entry in the cell cycle

  • An antibiotic often used as selection agent to determine transformation efficiency of algae, fungi, and mammalian cells
  • Serves as a selection agent for transformed cells carrying the antibiotic-resistance gene Sh ble
  • Causes DNA fragmentation by introducting double-strand breaks in DNA and prevents cells from entering S-phase during the cell cycle
TRUSTED_SUSTAINABILITY

Specifications

Type Phleomycin
Physical Form Liquid
Absorbance 0.75 to 1.05 AU at 244nm
CAS 11006-33-0
Source Streptomyces verticillus
Odor None
Packaging Plastic Bottle
Sterile Filtered Pass Test
Quantity 100 mg
Molecular Formula C51H75N17O21S2
InChI Key QRBLKGHRWFGINE-UGWAGOLRSA-N
SMILES CC1=C(N=C(N=C1N)C(CC(=O)N)NCC(C(=O)N)N)C(=O)NC(C(C2=CN=CN2)OC3C(C(C(C(O3)CO)O)O)(C)OC4C(C(C(C(O4)CO)O)(C(=O)N)O)O)C(=O)NC(C)C(C(C)C(=O)NC(C(=O)C)C(=O)NCCC5=NC(CS5)C6=NC(=CS6)C(=O)N)O
IUPAC Name 2-[2-[2-[[2-[[4-[[2-[[6-amino-2-[3-amino-1-[(2,3-diamino-3-oxopropyl)amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[(2R,3S,4S,5S,6S)-3-[(2S,3R,4R,5S)-4-carbamoyl-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)-
Molecular Weight (g/mol) 1326.379
PubChem CID 72511
Formula Weight 1526.5
Chemical Name or Material Phleomycin
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phleomycin, 2% solution in water
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H351-Suspected of causing cancer.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.