Learn More
Melford Phleomycin
Description
Phleomycin is an antibiotic agent. It is known for its ability to cleave DNA, making it useful for studies involving DNA damage and repair mechanisms.
Phleomycin, reconstituted, is often employed to induce controlled DNA damage in cells by causing single- and double-strand breaks in the DNA structure. Typically used to investigate cellular responses to DNA damage, as well as to explore the mechanisms involved in DNA repair.
It is a valuable tool for understanding various aspects of molecular biology, including gene regulation and genome integrity.
Specifications
Specifications
| Chemical Name or Material | Phleomycin |
| CAS | 11006-33-0 |
| Quantity | 25 mg |
| Molecular Formula | C55H85O21N20S2Cu-HCl |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.