Learn More
Melford Phleomycin 20 mg/mL Solution
Description
Phleomycin is an antibiotic and antitumor agent commonly used in molecular biology and life science research.
Phleomycin 20 mg/mL Solution is a formulation often employed in molecular biology experiments to induce controlled DNA damage in cells. Phleomycin accomplishes this by causing single- and double-strand breaks in DNA, making it a valuable tool for investigating cellular responses to DNA damage and the mechanisms involved in DNA repair.
Researchers may use Phleomycin 20 mg/mL Solution to study the impact of DNA damage on cellular processes, gene expression, and other aspects of molecular biology.
Specifications
Specifications
| Chemical Name or Material | Phleomycin 20 mg/ml Solution |
| CAS | 11006-33-0 |
| Quantity | 20 mg |
| Molecular Formula | C55H85O21N20S2Cu-HCl |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.