missing translation for 'onlineSavingsMsg'
Learn More

Melford Phleomycin 20 mg/mL Solution

Product Code. 30018652
Change view
Click to view available options
Quantity:
20 mg
100 mg
Unit Size:
100mg
20mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018652 20 mg 20mg
30018653 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018652 Supplier Melford Supplier No. P202200.02

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Phleomycin is an antibiotic and antitumor agent commonly used in molecular biology and life science research.

Phleomycin is an antibiotic and antitumor agent commonly used in molecular biology and life science research. It is known for its ability to cleave DNA, making it useful for studies involving DNA damage and repair mechanisms.

Phleomycin 20 mg/mL Solution is a formulation often employed in molecular biology experiments to induce controlled DNA damage in cells. Phleomycin accomplishes this by causing single- and double-strand breaks in DNA, making it a valuable tool for investigating cellular responses to DNA damage and the mechanisms involved in DNA repair.

Researchers may use Phleomycin 20 mg/mL Solution to study the impact of DNA damage on cellular processes, gene expression, and other aspects of molecular biology.

Specifications

Chemical Name or Material Phleomycin 20 mg/ml Solution
CAS 11006-33-0
Quantity 20 mg
Molecular Formula C55H85O21N20S2Cu-HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.