missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHLDB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557176
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Immunogen sequence: NPNTLKEGYI SVNEINEPCG NSTNLSPSTQ FPADADAVAT EPATAVLASQ PQSKEHFRSL EERKKQHKEG LYLSDTLPRK KTTSSISPHF SSATM Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
Seems to be involved in the assembly of the postsynaptic apparatus. May play a role in acetyl-choline receptor (AChR) aggregation in the postsynaptic membrane.
Tekniske data
| PHLDB2 | |
| Polyclonal | |
| Unconjugated | |
| PHLDB2 | |
| AV253284; C820004H04Rik; FLJ21791; LL5 beta; LL5b; LL5beta; Phldb2; pleckstrin homology like domain family B member 2; pleckstrin homology-like domain family B member 2; pleckstrin homology-like domain, family B, member 2; protein LL5-beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90102 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86SQ0 | |
| PHLDB2 | |
| Recombinant protein corresponding to Human PHLDB2. Recombinant protein control fragment (Product #RP-102653). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion