missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHLDA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584598
This item is not returnable.
View return policy
Description
Immunogen sequence: KGKYMYFTVV MAEGKEIDFR CPQDQGWNAE ITLQMVQYKN RQAILAVKST RQKQQHLVQQ QP Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 100%.
Cytotoxic T lymphocyte (CTL)-mediated cytotoxicity constitutes an important component of specific effector mechanisms in immunosurveillance against virus-infected or -transformed cells. Two mechanisms appear to account for this activity, one of which is the perforin-based process. Independently, a FAS-based mechanism involves the transducing molecule FAS (APO-1) and its ligand (FAS-L). The human FAS (APO-1) protein is a 48 kDa cell surface glycoprotein that belongs to a family of receptors that includes CD40, nerve growth factor receptors and tumor necrosis factor receptors. The FAS antigen is expressed on a broad range of lymphoid cell lines, and is expressed at high levels in T cells subsequent to crosslinking of the T cell receptor (TCR). A previously undescribed protein, TDAG51, restores activation- induced apoptosis in cells that have lost the ability to display Fas in response to activation.
Specifications
| PHLDA1 | |
| Polyclonal | |
| Unconjugated | |
| PHLDA1 | |
| apoptosis-associated nuclear protein; DT1P1B11; PHLDA1; PHRIP; pleckstrin homology like domain family A member 1; pleckstrin homology-like domain family A member 1; pleckstrin homology-like domain, family A, member 1; PQR protein; PQ-rich protein; proline- and glutamine-rich protein; Proline- and histidine-rich protein; proline-histidine rich protein; T-cell death associated; T-cell death-associated gene 51 protein; Tdag; TDAG51 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22822 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WV24 | |
| PHLDA1 | |
| Recombinant protein corresponding to Human PHLDA1. Recombinant protein control fragment (Product #RP-107243). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction