missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PHKG1, GST Tag Recombinant Protein

Product Code. 11523587
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11523587

Brand: Invitrogen™ PV3853

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

PHKG1 is a serine/threonine kinase with glycogenolytic regulatory functions. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. PHKG1 has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16816
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 5260
Molecular Weight (g/mol) 72.5 kDa
Name Human PHKG1, GST Tag
Quantity 10 μg
Regulatory Status For research use only. Not for use in diagnostic procedures
Gene Alias gamma phosphorylase kinase; MGC137450 protein; PHGK; PHKG; PHKG1; PHKgammaM; PHK-gammaM; PHK-gamma-M; PHKgamma-M; PHKIN01; phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform; Phosphorylase B kinase gamma catalytic chain, skeletal muscle isoform (Phosphorylase kinase gamma subunit 1); phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase; phosphorylase kinase catalytic subunit gamma 1; phosphorylase kinase gamma; phosphorylase kinase gamma 1; phosphorylase kinase gamma subunit; phosphorylase kinase gamma subunit 1; phosphorylase kinase subunit gamma 1; phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1; phosphorylase kinase, gamma 1 (muscle); serine/threonine-protein kinase PHKG1; unnamed protein product
Common Name PHKG1
KinaseFamily PHK
Product Type PHKG1 Recombinant Human Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Protein Family Kinase and inhibitors
Form Liquid
Protein Subtype Serine/threonine kinase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.