missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHKG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16743-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the Ser/Thr protein kinase family and encodes a protein with one protein kinase domain and two calmodulin-binding domains. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. This gene has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12.Specifications
| PHKG1 | |
| Polyclonal | |
| Unconjugated | |
| PHKG1 | |
| PHKG, PHKG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18682, 29353, 5260 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07934, P13286, Q16816 | |
| PHKG1 | |
| PHKG1 Fusion Protein Ag10032 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction