missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHKG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550728
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PHKG1 protein.
PHKG1 is a serine/threonine kinase with glycogenolytic regulatory functions. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. PHKG1 has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12.
Specifications
| PHKG1 | |
| Polyclonal | |
| Unconjugated | |
| PHKG1 | |
| gamma phosphorylase kinase; MGC137450 protein; PHGK; PHKG; PHKG1; PHKgammaM; PHK-gammaM; PHK-gamma-M; PHKgamma-M; PHKIN01; phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform; Phosphorylase B kinase gamma catalytic chain, skeletal muscle isoform (Phosphorylase kinase gamma subunit 1); phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase; phosphorylase kinase catalytic subunit gamma 1; phosphorylase kinase gamma; phosphorylase kinase gamma 1; phosphorylase kinase gamma subunit; phosphorylase kinase gamma subunit 1; phosphorylase kinase subunit gamma 1; phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1; phosphorylase kinase, gamma 1 (muscle); serine/threonine-protein kinase PHKG1; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18682, 5260 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P07934, Q16816 | |
| PHKG1 | |
| Synthetic peptide corresponding to a region derived from internal residues of human phosphorylase kinase, gamma 1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction