missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHKB Polyclonal Antibody
PHKB Polyclonal Antibody
Brand: Invitrogen PA567288
This item is not returnable.
View return policy
Description
PHKB Polyclonal Antibody for ICC/IF
Specifications
| PHKB | |
| Polyclonal | |
| Unconjugated | |
| PHKB | |
| AI463271; Phkb; Phosphorylase B; Phosphorylase b kinase regulatory subunit beta; phosphorylase kinase; phosphorylase kinase beta; phosphorylase kinase beta subunit; phosphorylase kinase beta-subunit; phosphorylase kinase regulatory subunit beta; phosphorylase kinase subunit beta; phosphorylase kinase, beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5257 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q93100 | |
| PHKB | |
| Recombinant Human PHKB. Recombinant protein control fragment (Product #RP-107926). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction