Learn More
Thermo Scientific™ Phire™ Hot Start DNA Polymerase
Product Code. 10237467 Shop All Thermo Scientific Products
Click to view available options
:
1000 Rxns (50uL vol)
Product Code. 10237467
Brand: Thermo Scientific™ F120L
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Phire Hot Start DNA Polymerase is the older version of Phire Hot Start DNA Polymerase; for upgraded version, please see Phire Hot Start II DNA Polymerase.
Thermo Scientific™ Phire™ Hot Start DNA Polymerase is the older version of this polymerase. For an upgraded version, please see Phire Hot Start II DNA Polymerase.
Note: Phire Hot-Start DNA Polymerase has been upgraded to new Phire Hot Start II DNA Polymerase (F-122S and F-122L). Current Phire Hot Start customers can switch to Phire Hot Start II with no changes needed to existing protocols or primers. Original Phire Hot Start will remain available for purchase, however, all new customers are advised to purchase Phire Hot Start II to allow more flexibility in primer design.
  • Quick hot start: No reactivation step
  • Fast enzyme: Amplify four times faster than with hot start Taq
  • Robust: Minimal reaction optimization due to high inhibitor tolerance
  • High yields: Abundant products due to high efficiency
  • Longer PCR products: Amplify significantly longer DNA fragments than with any hot start Taq
Includes:
Phire Hot Start DNA Polymerase is supplied with 5x Phire Reaction Buffer and DMSO. The buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Recommended for:
Fast PCR, routine PCR, high-throughput PCR, long-range PCR, DHPLC, microarray
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Phire Hot Start DNA Polymerase is supplied with 5x Phire Reaction Buffer and DMSO. The buffer provides 1.5 mM MgCl2 in the final 1x concentration. Recommended storage temperature -20°C. Stable for one year from the assay date.
Description Phire™ Hot Start DNA Polymerase
Quantity 1000 (50µL vol) reactions
For Use With (Application) Microarray Analysis