Learn More
Invitrogen™ PHIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110105
Description
Immunogen sequence: KTILENSVKH SKALNTLSSP GQSSFSHGTR NNSAKENMEK EKPVKRKMKS SVLPKASTLS KSSAVIEQGD CKNNALVPGT IQVNGHGGQP SKLVKRGPGR KPKVEVNTNS GEIIHKKRGR KPKKLQYAKP EDLEQNNVHP IRDEVLPSST CNFLSETNNV KEDLLQKKNR GGRKPKRKMK TQKLDADLLV PASVKVLRRS NRKKIDDPID EEEEFEELKG SEPHMRTRNQ GRRTAFYNED DSEEEQRQLL FEDTSLTFGT SSRGRVRKLT EKAKANLIGW.
PHIP (pleckstrin homology domain interacting protein), also known as ndrp or WDR11, is a 1,821 amino acid protein that contains eight N-terminal WD40 repeats and two bromodomains. It is expressed in skeletal muscle (localizing to the cytosol and nucleus) and primary beta cells (localizing to the nucleus) and acts as a transcriptional activator. PHIP is known to interact with various members of the insulin receptor substrate (IRS) family. The IRS family of proteins mediate insulin receptor signaling and play an important role in insulin-producing beta cell proliferation and survival. PHIP specifically associates with the PH domain of IRS-1 and may function to link IRS-1 to insulin receptors, indicating a vital role of PHIP in the regulation of insulin signaling.
Specifications
| PHIP | |
| Polyclonal | |
| Unconjugated | |
| PHIP | |
| 2810004D21Rik; 4632404O06Rik; BRWD2; DCAF14; DDB1 and CUL4 associated factor 14; DDB1- and CUL4-associated factor 14; FLJ20705; IRS-1 PH domain-binding protein; Ndrp; neuronal differentiation related protein; neuronal differentiation-related protein; PH-interacting protein; PHIP; pleckstrin homology domain interacting protein; WD repeat domain 11; WD repeat-containing protein 11; WDR11 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55023, 83946 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8VDD9, Q8WWQ0 | |
| PHIP | |
| Recombinant Protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.