missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-19096
This item is not returnable.
View return policy
Description
PHIP Polyclonal specifically detects PHIP in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| PHIP | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| DCAF14, DDB1 and CUL4 associated factor 14, FLJ20705, FLJ45918, IRS-1 PH domain-binding protein, MGC90216, ndrp, PH-interacting protein, pleckstrin homology domain interacting protein, WD repeat-containing protein 11, WDR11 | |
| Rabbit | |
| 207 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 55023 | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| Q8WWQ0 | |
| PHIP | |
| The immunogen recognized by this antibody maps to a region between residue 1771 and 1821 of human pleckstrin homology domain interacting protein using the numbering given in entry NP_060404.3 (GeneID 55023). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Zebrafish, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Green anole, White-tufted-ear marmoset, and Chinese hamster. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction