missing translation for 'onlineSavingsMsg'
Learn More

Phillygenin, MedChemExpress

Product Code. 16491418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491418 5 mg 5mg
16448998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491418 Supplier Medchem Express Supplier No. HYN0483
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phillygenin (Phillygenol) is an active ingredient from Forsythia with many medicinal properties, such as antioxidant, reducing blood lipid, inhibition of low density lipoprotein oxidation.

Phillygenin (Phillygenol) is an active ingredient from Forsythia with many medicinal properties, such as antioxidant, reducing blood lipid, inhibition of low density lipoprotein oxidation.

Specifications

Chemical Name or Material Phillygenin
CAS 487-39-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H24O6
Quantity 5 mg
Synonym Phillygenol Epipinoresinol methyl ether (+)-Phillygenin
Solubility Information DMSO : 50 mg/mL (134.26 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=CC=C([C@H]2OC[C@@]3([H])[C@]2([H])CO[C@H]3C4=CC=C(OC)C(OC)=C4)C=C1OC
Molecular Weight (g/mol) 372.41
Formula Weight 372.41
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.