missing translation for 'onlineSavingsMsg'
Learn More

Philanthotoxin 74 dihydrochloride, MedChemExpress

Product Code. 16453928
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
5 mg
10 mg
25 mg
missing translation for 'unitSize'
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16453928

missing translation for 'mfr': Medchem Express HY104020A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Philanthotoxin 74 dihydrochloride (PhTx 74) is an AMPAR antagonist; inhibits GluR3 and GluR1 with IC50s of 263 and 296 nM, respectively.

Philanthotoxin 74 dihydrochloride (PhTx 74) is an AMPAR antagonist

Specifications

Chemical Name or Material Philanthotoxin 74 dihydrochloride
CAS 1227301-51-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H44Cl2N4O3
Quantity 10 mg
Synonym PhTx 74 dihydrochloride
Solubility Information DMSO : 100 mg/mL (197.03 mM; Need ultrasonic) ∣H2O : 50 mg/mL (98.51 mM; Need ultrasonic)
SMILES O=C(NCCCCCCCNCCCCN)[C@@H](NC(CCC)=O)CC1=CC=C(O)C=C1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 507.54
Formula Weight 507.54
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.