missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PhiKan 083, Tocris Bioscience™
Description
p53 stabilizing agent; preferentially binds mutated (Y220C) p53 over wild-type p53 at a site distinct from functional DNA/protein interaction regions. Slows rate of thermal denaturation of p53-Y220C.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 274.79 |
| Percent Purity | >99% |
| Chemical Name or Material | PhiKan 083 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?