missing translation for 'onlineSavingsMsg'
Learn More

PhiKan 083, MedChemExpress

Product Code. 16446937
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16446937

Brand: Medchem Express HY108637

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PhiKan 083 is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM. PhiKan 083 can be used for cancer research.

PhiKan 083 is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM. PhiKan 083 can be used for cancer research.

Specifications

Chemical Name or Material PhiKan 083
CAS 880813-36-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H18N2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (419.59 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (419.59 mM; Need ultrasonic)
SMILES CNCC1=CC2=C(C=C1)N(CC)C3=C2C=CC=C3
Molecular Weight (g/mol) 238.33
Formula Weight 238.33
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.