missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PhiKan 083 hydrochloride, MedChemExpress
Description
PhiKan 083 hydrochloride is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM, and a relative binding affinity (Kd) of 150 μM in Ln229 cells.
Specifications
Specifications
| Chemical Name or Material | PhiKan 083 hydrochloride |
| CAS | 1050480-30-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H19ClN2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 62.5 mg/mL (227.45 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2 mg/mL (7.28 mM; Need ultrasonic and warming) |
| SMILES | CNCC1=CC2=C(C=C1)N(CC)C3=C2C=CC=C3.[H]Cl |
| Molecular Weight (g/mol) | 274.79 |
| Formula Weight | 274.79 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?