missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHGDH Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18850MFV610
This item is not returnable.
View return policy
Description
PHGDH Polyclonal antibody specifically detects PHGDH in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PHGDH | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| 3,3-phosphoglycerate dehydrogenase, 3PGDH, 3-PGDH, D-3-phosphoglycerate dehydrogenase, EC 1.1.1, EC 1.1.1.95, MGC3017, PDG, PGAD, PGD, PGDH, phosphoglycerate dehydrogenase, SERA | |
| Produced in rabbits immunized with E. coli-derived Human PHGDH fragment | |
| 0.1 mL | |
| Stem Cell Markers | |
| 26227 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction