missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHGDH Antibody (CL0555) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52939CL1
This item is not returnable.
View return policy
Description
PHGDH Monoclonal antibody specifically detects PHGDH in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| PHGDH | |
| Monoclonal | |
| CoraFluor 1 | |
| 3,3-phosphoglycerate dehydrogenase, 3PGDH, 3-PGDH, D-3-phosphoglycerate dehydrogenase, EC 1.1.1, EC 1.1.1.95, MGC3017, PDG, PGAD, PGD, PGDH, phosphoglycerate dehydrogenase, SERA | |
| This antibody was developed using a recombinant protein derived from O43175, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 26227 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0555 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction