missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHF8 Recombinant Rabbit Monoclonal Antibody (24GB945)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen PHF8
Clone 24GB945
Concentration 1 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275574
GREENER_CHOICE
View Documents
Invitrogen™
MA557153
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276120
View Documents
Invitrogen™
MA557154
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PHF8 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

PHD finger protein 8 is a jumonji domain containing protein. Like other members of the jumonji family, PHF8 may therefore play a role in histone demethylation. Mutations in PHF8 lead to Siderius type X-linked mental retardation (MRXSSD), a mild to borderline type of mental retardation.
Specifications

Specifications

PHF8
1 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q80TJ7, Q9UPP1
23133, 317425, 320595
A synthesized peptide derived from human PHF8 (aa 800-860).
Antibody
24GB945
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
PHF8
9830141C09Rik; histone lysine demethylase PHF8; JHDM1F; jumonji C domain-containing histone demethylase 1F; Kiaa1111; mKIAA1111; MRXSSD; PHD finger protein 8; PHF8; RGD1561065; ZNF422
PHF8
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.