missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF8 Rabbit anti-Human, Mouse, Rat, Clone: 2H9D4, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | PHF8 |
|---|---|
| Clone | 2H9D4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18358913
|
Bio-Techne
NBP3-15910-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347362
|
Novus Biologicals
NBP3-15910-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHF8 Monoclonal antibody specifically detects PHF8 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PHF8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| EC 1.14.11.27, histone lysine demethylase PHF8, jumonji C domain-containing histone demethylase 1F, KIAA1111ZNF422JHDM1F, MRXSSD, PHD finger protein 8DKFZp686E0868 | |
| A synthetic peptide corresponding to a sequence within amino acids 800-900 of human PHF8 (Q9UPP1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 2H9D4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 23133 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title