missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHF7 Monoclonal Antibody (1E9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521951
This item is not returnable.
View return policy
Description
Peptide Sequence: GRDSFEDEGR WCLILCATCG SHGTHRDCSS LRSNSKKWEC EECSPAAATD YIPENSGDIP CCSSTFHPEE HFCRDNTLEE NPGLSWTDWP EPSLLEKPES.
Spermatogenesis is a complex process regulated by extracellular and intracellular factors as well as cellular interactions among interstitial cells of the testis, Sertoli cells, and germ cells. In the testis, this gene is expressed in Sertoli cells but not germ cells. However, this gene is not expressed in a patient who exhibited spermatogenic arrest at the spermatocyte stage. Spermatogenic arrest is an interruption of germ cell differentiation that may result in oligospermia or azoospermia. The proteins encoded by this gene contain plant homeodomain (PHD) finger domains, also known as leukemia associated protein (LAP) domains, believed to be involved in transcriptional regulation. Thus this protein, which localizes to the nucleus of transfected cells, has been implicated in the transcriptional regulation of spermatogenesis. Two protein isoforms are encoded by transcript variants of this gene.
Specifications
| PHF7 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9BWX1 | |
| PHF7 | |
| PHF7 (NP_057567.3, 258 a.a. ~ 357 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1E9 | |
| Unconjugated | |
| PHF7 | |
| 1700006H01Rik; 1700010P14Rik; AI427892; AW555949; HSPC045; HSPC226; NYD-SP6; PHD finger protein 7; PHF7; testicular secretory protein Li 34; testis development protein NYD-SP6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51533 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction