missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF7 Antibody (OTI1G9), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73371H
This item is not returnable.
View return policy
Description
PHF7 Monoclonal specifically detects PHF7 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PHF7 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| PHF7 | |
| Recombinant protein fragment corresponding to amino acids 155-381 of human PHF7 (NP_057567) produced in E.coli. | |
| 0.1 mL | |
| 51533.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1G9 | |
| Western Blot, Immunohistochemistry | |
| DKFZp434L1850, HSPC045, HSPC226, MGC26088, NYD-SP6, PHD finger protein 7, Testis development protein NYD-SP6 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction