missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF5A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | PHF5A |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PHF5A Polyclonal antibody specifically detects PHF5A in Human, Rat samples. It is validated for Western BlotSpecifications
| PHF5A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 84844 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| bK223H9.2, INI, MGC1346, PHD finger protein 5A, PHD finger-like domain protein 5A, PHD finger-like domain-containing protein 5A, PHD-finger 5a, Rds3, SAP14b, SF3b14bSplicing factor 3B-associated 14 kDa protein, splicing factor 3B associated 14 kDa protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human PHF5A (NP_116147.1). MAKHHPDLIFCRKQAGVAIGRLCEKCDGKCVICDSYVRPCTLVRICDECNYGSYQGRCVICGGPGVSDAYYCKECTIQEKDRDGCPKIVNLGSSKTDLFYERKKYGFKKR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title