missing translation for 'onlineSavingsMsg'
Learn More

PHF23 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18431824 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431824 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431824 Supplier Novus Biologicals Supplier No. NBP128747AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PHF23 Polyclonal antibody specifically detects PHF23 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF23
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias FLJ16355, FLJ22884, hJUNE-1b, MGC2941, PDH-containing protein JUNE-1, PHD finger protein 23, PHD finger protein 23a
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 353 and 403 of human PHD finger protein 23 using the numbering given in entry NP_077273.2 (GeneID 79142).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79142
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.