missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF23 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28746AF350
This item is not returnable.
View return policy
Description
PHF23 Polyclonal antibody specifically detects PHF23 in Human samples. It is validated for Western Blot
Specifications
| PHF23 | |
| Polyclonal | |
| Western Blot | |
| FLJ16355, FLJ22884, hJUNE-1b, MGC2941, PDH-containing protein JUNE-1, PHD finger protein 23, PHD finger protein 23a | |
| The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human PHD finger protein 23 using the numbering given in entry NP_077273.2 (GeneID 79142). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79142 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction