Learn More
Invitrogen™ PHF22 Recombinant Rabbit Monoclonal Antibody (2D3T4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557956
Description
Immunogen sequence: MAATVNLELD PIFLKALGFL HSKSKDSAEK LKALLDESLA RGIDSSYRPS QKDVEPPKIS STKNISIKQE PKISSSLPSG NNNGKVLTTE KVKKEAEKRP ADKMKSDITE GVDIPKKPRL EKPETQSSPI TVQSSKDLPM ADLSSFEETS ADDFAMEMGL ACVVCRQMMV ASGNQLVECQ ECHNLYHRDC HKPQVTDKEA NDPRLVWYCA RCTRQMKRMA QKTQKPPQKP.
INTS12 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit and mediates 3-prime end processing of small nuclear RNAs U1 and U2.
Specifications
| PHF22 | |
| Recombinant Monoclonal | |
| 0.58 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9D168 | |
| Ints12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human INTS12 (NP_065128.2). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 2D3T4 | |
| Unconjugated | |
| Ints12 | |
| 1110020M19Rik; 2810027J24Rik; 4930529N21Rik; A230056J18Rik; AI666778; hypothetical nuclear factor SBBI22; hypothetical protein LOC415251; int12; Integrator complex subunit 12; INTS12; PHD finger protein 22; PHF22; SBBI22; Unknown (protein for MGC:134302); wu:fi17f05; zgc:86602 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 71793 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.