missing translation for 'onlineSavingsMsg'
Learn More

PHF21A Antibody, DyLight 755, Novus Biologicals™

Product Code. 18402974 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402974 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402974 Supplier Novus Biologicals Supplier No. NBP204043IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PHF21A Polyclonal antibody specifically detects PHF21A in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF21A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias BHC80a, BHC80BM-006, BRAF35-HDAC complex protein BHC80, KIAA1696BRAF35/HDAC2 complex (80 kDa), PHD finger protein 21A
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to this antibody using the numbering given in entry NP_001095272.1 (GeneID 51317).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 51317
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.