missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF20L1 Antibody (OTI6G6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45020-100ul
This item is not returnable.
View return policy
Description
PHF20L1 Monoclonal antibody specifically detects PHF20L1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| PHF20L1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 51105 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI6G6 | |
| Western Blot 1:500-2000, Immunohistochemistry 1:500 | |
| CGI-72, FLJ13649, FLJ21615, MGC64923, PHD finger protein 20-like 1, PHD finger protein 20-like protein 1, tudor domain-containing protein PHF20L1 | |
| Full length human recombinant protein of human PHF20L1 (NP_940915) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction