missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF20L1 Antibody (OTI1H6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | PHF20L1 |
|---|---|
| Clone | OTI1H6 |
| Dilution | Immunohistochemistry 1:500 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306261
|
Novus Biologicals
NBP3-45019-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306179
|
Novus Biologicals
NBP3-45019-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHF20L1 Monoclonal antibody specifically detects PHF20L1 in Human,Mouse,Rat samples. It is validated for ImmunohistochemistrySpecifications
| PHF20L1 | |
| Immunohistochemistry 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 51105 | |
| IgG1 | |
| >95% | |
| Antibody |
| OTI1H6 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| CGI-72, FLJ13649, FLJ21615, MGC64923, PHD finger protein 20-like 1, PHD finger protein 20-like protein 1, tudor domain-containing protein PHF20L1 | |
| Full length human recombinant protein of human PHF20L1 (NP_940915) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title