missing translation for 'onlineSavingsMsg'
Learn More

PHF20L1 Antibody (OTI1B4) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30688665 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688665 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688665 Supplier Novus Biologicals Supplier No. NBP345018MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PHF20L1 Monoclonal antibody specifically detects PHF20L1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF20L1
Applications Western Blot
Classification Monoclonal
Clone OTI1B4
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias CGI-72, FLJ13649, FLJ21615, MGC64923, PHD finger protein 20-like 1, PHD finger protein 20-like protein 1, tudor domain-containing protein PHF20L1
Host Species Mouse
Immunogen Full length human recombinant protein of human PHF20L1 (NP_940915) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51105
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.