Learn More
Invitrogen™ PHF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14120
Description
PA1-4120 detects PHF-2 in human samples. PA1-4120 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~120 kDa protein representing PHF-2 in human placenta lysate. The PA1-4120 immunogen is a synthetic peptide corresponding to residues K(70) H G P G P T P D V K R V C(82) of the 120 kDa PHF.
This gene encodes a protein which contains a zinc finger-like PHD (plant homeodomain) finger, distinct from other classes of zinc finger motifs, and a hydrophobic and highly conserved domain. The PHD finger shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure.
Specifications
| PHF2 | |
| Polyclonal | |
| Unconjugated | |
| PHF2 | |
| CENP-35; centromere protein 35; GRC5; JHDM1E; jumonji C domain-containing histone demethylase 1E; Kiaa0662; Lysine-specific demethylase PHF2; PHD finger protein 2; Phf2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5253 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O75151 | |
| PHF2 | |
| Synthetic peptide corresponding to residues K(70) H G P G P T P D V K R V C(82) of the 120 kDa PHF. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.