missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHF13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524986
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology.
PHF13 contains 1 PHD-type zinc finger. The function of PHF13 is currently unknown.
Specifications
| PHF13 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q86YI8, Q8K2W6 | |
| PHF13 | |
| KLH conjugated synthetic peptide between 263-291 amino acids from the C-terminal region of human PHF13. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Hamster | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| PHF13 | |
| BC029632; PHD finger protein 13; PHD zinc finger protein PHF5; PHF13; PHF5; SPOC1; Survival time-associated PHD finger protein in ovarian cancer 1; survival time-associated PHD protein in ovarian cancer | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100760522, 148479, 230936 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction