missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97688-100ul
This item is not returnable.
View return policy
Description
PHF13 Polyclonal antibody specifically detects PHF13 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PHF13 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| MGC43399, PHD finger protein 13, PHD zinc finger protein PHF5, PHF5, SPOC1, Survival time-associated PHD finger protein in ovarian cancer 1, survival time-associated PHD protein in ovarian cancer | |
| Produced in rabbits immunized with E. coli-derived Human PHF13 fragment. | |
| 100 μg | |
| Cell Biology | |
| 148479 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction