missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHF12 Recombinant Mouse Monoclonal Antibody (K8U017_20C4)
GREENER_CHOICE

Product Code. 30829308
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829308 100 μL
30825071 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829308 Supplier Invitrogen™ Supplier No. MA564424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: RSASKTELKA IAHARILERR ASRPGTPTSS ASTETPTSEQ NDVDEDIIDV DEEPVAAEPD YVQPQLRRPF ELLIAAAMER NPTQFQLPNE LTCTTALPGS SKRRRKEETT GKNVKKTQHE LDHNGLVPLP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Pig - 99%, Cynomolgus monkey - 99%.

Acts as a transcriptional repressor. Involved in recruitment of functional SIN3A complexes to DNA. Represses transcription at least in part through the activity of an associated histone deacetylase (HDAC). May also repress transcription in a SIN3A-independent manner through recruitment of functional TLE5 complexes to DNA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF12
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K8U017_20C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene PHF12
Gene Accession No. Q96QT6
Gene Alias 2410142K10Rik; F630045O13; KIAA1523; mKIAA1523; Pf1; PHD factor 1; PHD finger protein 12; PHD zinc finger transcription factor; phf12; PHF12 (HGNC:20816)
Gene Symbols PHF12
Host Species Mouse
Immunogen Recombinant human PHF12 (aa 141-270).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57649
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.