missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10898-1-AP-20UL
This item is not returnable.
View return policy
Description
PHF11 contains two PHD zinc fingers and probably regulates transcription. Distinctive splice variants were expressed in immune tissues and cells.Specifications
| PHF11 | |
| Polyclonal | |
| Unconjugated | |
| PHF11 | |
| APY, IGEL, IGER, IGHER, NY REN 34, NYREN34, PHD finger protein 11, PHF11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 219132, 51131 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A6H5X4, Q9UIL8 | |
| PHF11 | |
| PHF11 Fusion Protein Ag1318 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction