missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHF10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112648
This item is not returnable.
View return policy
Description
PHF10 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This gene contains a predicted ORF that encodes a protein with two zinc finger domains. The function of the encoded protein is not known. Sequence analysis suggests that multiple alternatively spliced transcript variants are derived from this gene but the full-length nature of only two of them is known. These two splice variants encode different isoforms. A pseudogene for this gene is located on Xq28.
Specifications
| PHF10 | |
| Polyclonal | |
| Unconjugated | |
| PHF10 | |
| 1810055P05Rik; AV024533; BAF45A; BRG1-associated factor 45a; BRG1-associated factor, 45-KD, A; PHD finger protein 10; PHD finger protein 10, isoform a; PHD zinc finger protein XAP135; Phf10; XAP135 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 55274 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8WUB8 | |
| PHF10 | |
| Recombinant Human PHD finger protein 10 protein (151-360AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction