missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF10 Antibody (PCRP-PHF10-2A10), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14153UV
This item is not returnable.
View return policy
Description
PHF10 Monoclonal antibody specifically detects PHF10 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| PHF10 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 55274 | |
| Store at 4°C in the dark. | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-PHF10-2A10 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| BAF45a, BAF45APHD zinc finger protein XAP135, BRG1-associated factor 45a, FLJ10975, MGC111009, PHD finger protein 10, XAP135BRG1-associated factor, 45-KD, A | |
| Recombinant full-length humanPHF10protein (Uniprot: Q8WUB8) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction