missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94819-0.02ml
This item is not returnable.
View return policy
Description
PHF10 Polyclonal antibody specifically detects PHF10 in Human, Mouse samples. It is validated for Western Blot
Specifications
| PHF10 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| BAF45a, BAF45APHD zinc finger protein XAP135, BRG1-associated factor 45a, FLJ10975, MGC111009, PHD finger protein 10, XAP135BRG1-associated factor, 45-KD, A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PHF10 (NP_060758.2). MAAAAGPGAALSPRPCDSDPATPGAQSPKDDNEDNSNDGTQPSKRRRMGSGDSSRSCETSSQDLGFSYYPAENLIEYKWPPDETGEYYMLQEQVSEYLGVTSFKRKYPDLERRDLSHKEKLYLRELNVITETQCTLGLTALRSDEVIDLMIKEYPAKHAEYSVILQEKERQRITDHYKEYSQMQQQNTQKVEASKVPEYIKKAAKKAAEFNSNLNRERMEERRAYFDLQTHVIQVPQGKYKVLPTERTKVSSYPVALIPGQFQEYYKRYSPDELRYLPLNTALYEPPLDPELPALDSDGD | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55274 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction