missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ pHep™ Portable pH/Temperature Meter
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
Description
- Waterproof pH meters, choice of models 0.1/0.01pH
- Large, easy to read dual LCD showing pH and °C
- Replaceable electrode
- Automatic temperature compensation
- Automatic calibration, one or two point with choice of five buffer values
- Hold function and automatic switch off
- Floats if accidentally dropped in water
- Includes a protective cap and non-clogging fibre reference junction, which can be replaced by simply plugging in a new one
Specifications
Specifications
| Accuracy (Temperature) | ±0.5°C/±1°F |
| Accuracy (pH) | ±0.1 |
| For Use With (Application) | pH calibration |
| Range (pH) | -2 to 16 |
| Resolution (pH) | 0.1 |
| Calibration Points | 4.01, 7.01, 10.01 or 4.01, 6.86, 9.18 |
| Temperature Range (Metric) | -5.0°C to +60.0°C |
| Temperature Range (English) | 23°F to 140°F |
| Resolution (Temperature) | 0.1°C, 0.1°F |
| Display Type | LCD |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction