missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ pHep™ Portable pH/Temperature Meter

Product Code. 10560133
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10560133

Brand: Hanna Instruments™ HI98128

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Features automatic one or two-point calibration and automatic temperature compensation. Hanna Instruments™ pHep™ Portable pH/Temperature Meter contains many features found in more expensive portable and benchtop meters.

  • Waterproof pH meters, choice of models 0.1/0.01pH
  • Large, easy to read dual LCD showing pH and °C
  • Replaceable electrode
  • Automatic temperature compensation
  • Automatic calibration, one or two point with choice of five buffer values
  • Hold function and automatic switch off
  • Floats if accidentally dropped in water
  • Includes a protective cap and non-clogging fibre reference junction, which can be replaced by simply plugging in a new one

Specifications

Accuracy (Temperature) ±0.5°C/±1°F
Accuracy (pH) ±0.05
For Use With (Application) pH calibration
Range (pH) -2 to 16
Resolution (pH) 0.01
Calibration Points 4.01, 7.01, 10.01 or 4.01, 6.86, 9.18
Temperature Range (Metric) -5.0°C to +60.0°C
Temperature Range (English) 23°F to 140°F
Resolution (Temperature) 0.1°C, 0.1°F
Display Type LCD
Weight (Metric) 100 g
Warranty 1 Year (Limited Warranty) Meter and 6 Month (Limited Warranty) Electrode
Material Rubberized Body
Model HI98128
Power Supply Yes
Probe Type Replaceable
Detection Range -2.0 to 16.0 pH, 1.5 V, -5.0°C to +60.0°C/23.0°F to 140.0°F Temperature
Temperature Compensation Automatic
Battery Type 1.5V
Type Waterproof Portable pH Meter
Battery Life 300 hr.
Voltage 1.5 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.