missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Pheos B-WR Safety Helmet
Shop All Uvex Products
Click to view available options
Color:
Black
White
Unit Size:
Each
Description
- Resistant to hot molten metal splash and temperatures down to -30°C (EN 367 additional requirement MM)
- Side euroslot adapters (30 mm) are suitable for the addition of earmuffs or the uvex pheos visor system
- Suspension harness with wheel ratchet allows continuous width adjustment, for a perfect fit at all times
- Six-point textile band attachment ensures optimal fit and comfort
- Available in a wide variety of color options
Specifications
Specifications
| Color | White |
| Model | Pheos B-WR |
| Ventilated | Ventilated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction