missing translation for 'onlineSavingsMsg'
Learn More

Phenytoin sodium, MedChemExpress

Product Code. 16450938
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16450938

Brand: Medchem Express HYB0448A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phenytoin sodium (5,5-Diphenylhydantoin sodium salt) is a potent Voltage-gated Na+ channels (VGSCs) blocker. Phenytoin has antiepileptic activity and reduces breast tumour growth and metastasis in mice.

Phenytoin sodium (5,5-Diphenylhydantoin sodium salt) is a potent Voltage-gated Na+ channels (VGSCs) blocker. Phenytoin has antiepileptic activity and reduces breast tumour growth and metastasis in mice.

Specifications

Chemical Name or Material Phenytoin sodium
Physical Form Solid
CAS 630-93-3
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C15H11N2NaO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym 5,5-Diphenylhydantoin sodium salt
Solubility Information DMSO : 50 mg/mL (182.32 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.65 mM; Need ultrasonic)
SMILES O=C1N=C(O[Na])NC1(C2=CC=CC=C2)C3=CC=CC=C3
Molecular Weight (g/mol) 274.25
Formula Weight 274.25
Percent Purity 99.96%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.