missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylammonium chloride, 98+%, ACROS Organics™

Product Code. 10773381 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10773381 100g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10773381 Supplier Thermo Scientific Acros Supplier No. 189791000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Denna artikel kan inte returneras. Se returpolicy
Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifikationer

CAS 138-24-9
Color Blue-Gray or White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Molecular Formula C9H14ClN
Linear Formula C6H5N(CH3)3Cl
MDL Number MFCD00011790
Quantity 100 g
Beilstein 12, 158
Synonym phenyltrimethylammonium chloride, n,n,n-trimethylbenzenaminium chloride, trimethylphenylammonium chloride, n,n,n-trimethylanilinium chloride, ammonyx 200, benzenaminium, n,n,n-trimethyl-, chloride, trimethylphenylammoniumchloride, trimethylanilinium chloride, ammonium, phenyltrimethyl-, chloride, trimethylphenyl ammonium chloride
InChI Key MQAYPFVXSPHGJM-UHFFFAOYSA-M
SMILES [Cl-].C[N+](C)(C)C1=CC=CC=C1
IUPAC Name N,N,N-trimethylanilinium chloride
Molecular Weight (g/mol) 171.67
PubChem CID 67309
Formula Weight 171.67
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material Phenyltrimethylammonium chloride, 98+%
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.