missing translation for 'onlineSavingsMsg'
Learn More

Phenyltriethoxysilane, 98%

Product Code. 11341667
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11341667 250 g 250g
11331667 50 g 50g
11351667 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11341667 Supplier Thermo Scientific Alfa Aesar Supplier No. L04684.30
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 780-69-8 | C12H20O3Si | 240.37 g/mol

Phenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 780-69-8
Molecular Formula C12H20O3Si
Molecular Weight (g/mol) 240.37
MDL Number MFCD00009065
InChI Key JCVQKRGIASEUKR-UHFFFAOYSA-N
Synonym phenyltriethoxysilane, triethoxy phenyl silane, silane, triethoxyphenyl, triethoxyfenylsilan, silane, phenyltriethoxy, benzeneorthosiliconic acid, triethyl ester, phenyl triethoxysilane, benzene, triethoxysilyl, triethoxyfenylsilan czech, unii-qi310t2x15
PubChem CID 13075
IUPAC Name triethoxy(phenyl)silane
SMILES CCO[Si](OCC)(OCC)C1=CC=CC=C1

Specifications

Boiling Point 235°C
Quantity 250 g
Flash Point 96°C (204°F)
Refractive Index 1.461
Sensitivity Moisture sensitive
Density 0.996
Linear Formula C6H5Si(OCH2CH3)3
Beilstein 2940602
Solubility Information Hydrolyzes in water.
Formula Weight 240.38
Percent Purity 98%
Chemical Name or Material Phenyltriethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.