missing translation for 'onlineSavingsMsg'
Learn More

Phenyltin trichloride

Product Code. 11354849
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354849 5 g 5g
11364849 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354849 Supplier Thermo Scientific Alfa Aesar Supplier No. 071157.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1124-19-2 | C6H5Cl3Sn | 302.166 g/mol

Phenyltin trichloride is used to produce 3-phenyl-cyclohexanone with cyclohex-2-enone. And it is also used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-19-2
Molecular Formula C6H5Cl3Sn
Molecular Weight (g/mol) 302.166
MDL Number MFCD00000513
InChI Key UBOGEXSQACVGEC-UHFFFAOYSA-K
Synonym phenyltin trichloride, phenyltrichlorotin, stannane, trichlorophenyl, trichlorophenyltin, trichloro phenyl stannane, phenyltrichlorostannane, ccris 6323, phsncl3, phenyl tin trichloride, monophenyltin trichloride
PubChem CID 14298
IUPAC Name trichloro(phenyl)stannane
SMILES C1=CC=C(C=C1)[Sn](Cl)(Cl)Cl

Specifications

Density 1.84 g/mL
Odor Pungent
Quantity 5 g
UN Number UN2788
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 302.18
Physical Form Liquid
Chemical Name or Material Phenyltin trichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.