missing translation for 'onlineSavingsMsg'
Learn More

Phenyltin Trichloride, TRC

Product Code. 30419947
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419947 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419947 Supplier Toronto Research Chemicals Supplier No. TRCP3996031G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyltin Trichloride
CAS 1124-19-2
Molecular Formula C6 H5 Cl3 Sn
Quantity 1 g
Synonym Phenyltin trichloride (6CI), Trichlorophenylstannane, Monophenyltin chloride, Monophenyltin trichloride, NSC 92617, Phenyltrichlorostannane, Phenyltrichlorotin, Trichlorophenyltin
InChI Formula InChI=1S/C6H5.3ClH.Sn/c1-2-4-6-5-3-1;;;;/h1-5H;3*1H;/q;;;;+3/p-3
SMILES Cl[Sn](Cl)(Cl)c1ccccc1
IUPAC Name trichloro(phenyl)stannane
Molecular Weight (g/mol) 302.17
Formula Weight 301.8479
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.